Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLVS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319392.1 | NP_001306321.1 | 4 - 5 | 985 | 84 | |
XM_005551715.2 | XP_005551772.1 | 4 - 5 | 2589 | 84 | ||
GenBank mRNA | AB060190.1 | - | 3 - 4 | 1001 | 84 | |
AB060902.1 | - | 4 - 5 | 985 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |