Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319585.1 | NP_001306514.1 | 3 - 4 | 1671 | 115 | |
XM_005561890.2 | XP_005561947.1 | 2 - 3 | 1792 | 115 | ||
GenBank mRNA | AB070164.1 | - | 3 - 4 | 1671 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |