Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549563.2 | XP_005549620.1 | 3 - 4 | 317 | 150 | |
XM_005549564.1 | XP_005549621.1 | 2 - 3 | 228 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |