Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102143414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015434074.1 | XP_015289560.1 | 5 - 6 | 663 | 97 | |
XM_015434075.1 | XP_015289561.1 | 5 - 6 | 561 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |