Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582858.2 | XP_005582915.1 | 1 | 954 | 117 | |
XM_005582859.2 | XP_005582916.1 | 3 | 447 | 117 | ||
XM_015437629.1 | XP_015293115.1 | 2 | 880 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |