Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015441596.1 | XP_015297082.1 | 10 - 11 | 2113 | 87 | |
XM_015441597.1 | XP_015297083.1 | 10 - 11 | 2157 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |