Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549343.2 | XP_005549400.1 | 3 - 4 | 440 | 101 | |
XM_015447207.1 | XP_015302693.1 | 3 - 4 | 584 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Inventoried Probe spans exons |