Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015455389.1 | XP_015310875.1 | 3 - 4 | 1585 | 90 | |
XM_015455390.1 | XP_015310876.1 | 2 - 3 | 1571 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |