Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOX2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005554779.2 | XP_005554836.1 | 2 - 3 | 635 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |