Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015438389.1 | XP_015293875.1 | 6 - 7 | 1462 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |