Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL36A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015433412.1 | XP_015288898.1 | 3 - 4 | 682 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |