Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS37C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577684.2 | XP_005577741.1 | 5 - 6 | 576 | 65 | |
XM_005577685.2 | XP_005577742.1 | 4 - 5 | 641 | 65 | ||
XM_005577686.2 | XP_005577743.1 | 4 - 5 | 1985 | 65 | ||
XM_015434644.1 | XP_015290130.1 | 3 - 4 | 1015 | 65 | ||
XM_015434645.1 | XP_015290131.1 | 4 - 5 | 643 | 65 | ||
XM_015434646.1 | XP_015290132.1 | 5 - 6 | 2042 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |