Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579132.2 | XP_005579189.1 | 8 - 9 | 1114 | 110 | |
XM_005579133.2 | XP_005579190.1 | 8 - 9 | 1111 | 110 | ||
XM_005579134.2 | XP_005579191.1 | 7 - 8 | 995 | 110 | ||
XM_005579135.2 | XP_005579192.1 | 8 - 9 | 1114 | 110 | ||
XM_005579136.2 | XP_005579193.1 | 7 - 8 | 992 | 110 | ||
XM_005579137.2 | XP_005579194.1 | 7 - 8 | 961 | 110 | ||
XM_005579138.2 | XP_005579195.1 | 8 - 9 | 1114 | 110 | ||
XM_005579139.2 | XP_005579196.1 | 7 - 8 | 961 | 110 | ||
XM_015435510.1 | XP_015290996.1 | 6 - 7 | 841 | 110 | ||
XM_015435511.1 | XP_015290997.1 | 8 - 9 | 1114 | 110 | ||
XM_015435512.1 | XP_015290998.1 | 7 - 8 | 995 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |