Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591033.2 | XP_005591090.1 | 3 - 4 | 903 | 114 | |
XM_015442612.1 | XP_015298098.1 | 3 - 4 | 472 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |