Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284093.1 | NP_001271022.1 | 2 - 3 | 196 | 149 | |
GenBank mRNA | AB169713.1 | - | 2 - 3 | 201 | 149 | |
AB170858.1 | - | 2 - 3 | 196 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |