Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JADE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555911.2 | XP_005555968.1 | 8 - 9 | 1121 | 93 | |
XM_005555912.2 | XP_005555969.1 | 8 - 9 | 1310 | 93 | ||
XM_005555913.2 | XP_005555970.1 | 8 - 9 | 1261 | 93 | ||
XM_005555914.2 | XP_005555971.1 | 8 - 9 | 1132 | 93 | ||
XM_015450829.1 | XP_015306315.1 | 8 - 9 | 1137 | 93 | ||
XM_015450830.1 | XP_015306316.1 | 8 - 9 | 1134 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |