Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285245.1 | NP_001272174.1 | 2 - 3 | 246 | 81 | |
XM_005560075.2 | XP_005560132.1 | 2 - 3 | 728 | 81 | ||
XM_005560076.2 | XP_005560133.1 | 3 - 4 | 466 | 81 | ||
XM_015452452.1 | XP_015307938.1 | 3 - 4 | 482 | 81 | ||
XM_015452453.1 | XP_015307939.1 | 3 - 4 | 407 | 81 | ||
GenBank mRNA | AB168338.1 | - | 2 - 3 | 246 | 81 |