Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005550928.2 | XP_005550985.1 | 4 - 5 | 1264 | 72 | |
XM_005550929.2 | XP_005550986.1 | 4 - 5 | 386 | 72 | ||
XM_015448136.1 | XP_015303622.1 | 4 - 5 | 493 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |