Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576206.2 | XP_005576263.1 | 2 - 3 | 409 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |