Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575103.2 | XP_005575160.1 | 1 - 2 | 566 | 109 | |
XM_005575104.2 | XP_005575161.1 | 1 - 2 | 566 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |