Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283096.1 | NP_001270025.1 | 10 - 11 | 1852 | 92 | |
XM_005550638.2 | XP_005550695.1 | 11 - 12 | 2381 | 92 | ||
XM_005550639.2 | XP_005550696.1 | 9 - 10 | 2182 | 92 | ||
XM_005550640.2 | XP_005550697.1 | 9 - 10 | 2179 | 92 | ||
XM_015447303.1 | XP_015302789.1 | 10 - 11 | 1787 | 92 | ||
GenBank mRNA | AB169093.1 | - | 10 - 11 | 1852 | 92 | |
AB169129.1 | - | 8 - 9 | 1249 | 92 |