Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589807.2 | XP_005589864.1 | 3 - 4 | 1542 | 139 | |
XM_015441771.1 | XP_015297257.1 | 1 - 2 | 391 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |