Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015441154.1 | XP_015296640.1 | 5 - 6 | 2543 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |