Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005573439.2 | XP_005573496.1 | 1 - 2 | 520 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |