Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005550869.2 | XP_005550926.1 | 1 - 2 | 669 | 80 | |
XM_005550871.2 | XP_005550928.1 | 1 - 2 | 669 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Probe spans exons |