Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005571048.2 | XP_005571105.1 | 1 - 2 | 802 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |