Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563671.2 | XP_005563728.1 | 2 - 3 | 998 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |