Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102130411 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005584176.1 | XP_005584233.1 | 1 | 169 | 174 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Both primers and probe map within a single exon |