Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553771.2 | XP_005553828.1 | 1 | 275 | 81 | |
XM_005553772.2 | XP_005553829.1 | 1 | 128 | 81 | ||
XM_005553773.2 | XP_005553830.1 | 1 | 278 | 81 | ||
XM_005553774.2 | XP_005553831.1 | 1 | 270 | 81 | ||
XM_005553775.2 | XP_005553832.1 | 1 | 256 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Both primers and probe map within a single exon |