Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287637.1 | NP_001274566.1 | 4 - 5 | 650 | 81 | |
XM_005589322.2 | XP_005589379.1 | 4 - 5 | 697 | 81 | ||
GenBank mRNA | DQ074793.1 | - | 4 - 5 | 650 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Inventoried Probe spans exons |