Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005574247.2 | XP_005574304.1 | 3 | 429 | 110 | |
XM_005574248.2 | XP_005574305.1 | 4 | 521 | 110 | ||
XM_005574249.2 | XP_005574306.1 | 2 | 354 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Inventoried Both primers and probe map within a single exon |