Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015448297.1 | XP_015303783.1 | 1 - 2 | 590 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |