Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575655.2 | XP_005575712.1 | 4 - 5 | 400 | 80 | |
XM_005575656.2 | XP_005575713.1 | 3 - 4 | 294 | 80 | ||
XM_015433567.1 | XP_015289053.1 | 2 - 3 | 232 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |