Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102117337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569119.2 | XP_005569176.1 | 3 - 4 | 451 | 57 | |
XM_015429878.1 | XP_015285364.1 | 2 - 3 | 886 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |