Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589349.2 | XP_005589406.1 | 3 - 4 | 1129 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |