Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544773.2 | XP_005544830.1 | 1 - 2 | 323 | 76 | |
XM_005544774.2 | XP_005544831.1 | 3 - 4 | 344 | 76 | ||
XM_005544775.2 | XP_005544832.1 | 2 - 3 | 207 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |