Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53INP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563744.2 | XP_005563801.1 | 5 - 6 | 1958 | 94 | |
XM_005563745.2 | XP_005563802.1 | 4 - 5 | 1793 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |