Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576698.2 | XP_005576755.1 | 2 - 3 | 1464 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |