Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576065.2 | XP_005576122.1 | 4 - 5 | 441 | 102 | |
XM_005576066.2 | XP_005576123.1 | 5 - 6 | 474 | 102 | ||
XM_005576068.2 | XP_005576125.1 | 4 - 5 | 502 | 102 | ||
XM_005576069.2 | XP_005576126.1 | 3 - 4 | 365 | 102 | ||
XM_015433769.1 | XP_015289255.1 | 5 - 6 | 373 | 102 | ||
XM_015433770.1 | XP_015289256.1 | 5 - 6 | 374 | 102 | ||
XM_015433771.1 | XP_015289257.1 | 4 - 5 | 517 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |