Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549946.2 | XP_005550003.1 | 5 - 6 | 564 | 68 | |
XM_005549947.2 | XP_005550004.1 | 4 - 5 | 512 | 68 | ||
XM_015447621.1 | XP_015303107.1 | 5 - 6 | 590 | 68 | ||
XM_015447622.1 | XP_015303108.1 | 5 - 6 | 1072 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |