Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285236.1 | NP_001272165.1 | 4 - 5 | 699 | 107 | |
XM_005593053.2 | XP_005593110.1 | 4 - 5 | 847 | 107 | ||
GenBank mRNA | AB179464.1 | - | 4 - 5 | 699 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |