Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285236.1 | NP_001272165.1 | 7 - 8 | 1105 | 100 | |
XM_005593053.2 | XP_005593110.1 | 7 - 8 | 1253 | 100 | ||
GenBank mRNA | AB179464.1 | - | 7 - 8 | 1105 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |