Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005560108.2 | XP_005560165.1 | 1 | 1491 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |