Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283167.1 | NP_001270096.1 | 4 - 5 | 411 | 66 | |
GenBank mRNA | EF538683.1 | - | 4 - 5 | 411 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |