Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283742.1 | NP_001270671.1 | 3 - 4 | 476 | 104 | |
XM_005563807.2 | XP_005563864.1 | 4 - 5 | 584 | 104 | ||
XM_005563808.2 | XP_005563865.1 | 3 - 4 | 728 | 104 | ||
XM_005563809.2 | XP_005563866.1 | 3 - 4 | 527 | 104 | ||
XM_005563810.2 | XP_005563867.1 | 4 - 5 | 585 | 104 | ||
XM_015454805.1 | XP_015310291.1 | 4 - 5 | 798 | 104 | ||
XM_015454806.1 | XP_015310292.1 | 3 - 4 | 991 | 104 | ||
GenBank mRNA | AB172661.1 | - | 3 - 4 | 476 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |