Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284540.1 | NP_001271469.1 | 4 - 5 | 557 | 110 | |
XM_015430176.1 | XP_015285662.1 | 7 - 8 | 824 | 110 | ||
GenBank mRNA | JX866725.1 | - | 4 - 5 | 557 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |