Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284779.1 | NP_001271708.1 | 6 - 7 | 905 | 81 | |
GenBank mRNA | AB169626.1 | - | 6 - 7 | 905 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |