Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN1SW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283204.1 | NP_001270133.1 | 2 - 3 | 528 | 84 | |
GenBank mRNA | AF158977.1 | - | 2 - 3 | 528 | 84 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Probe spans exons |