Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283720.1 | NP_001270649.1 | 1 - 2 | 304 | 99 | |
GenBank mRNA | AB168990.1 | - | 1 - 2 | 304 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |