Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285355.1 | NP_001272284.1 | 6 - 7 | 705 | 76 | |
XM_015447283.1 | XP_015302769.1 | 5 - 6 | 649 | 76 | ||
XM_015447284.1 | XP_015302770.1 | 5 - 6 | 1891 | 76 | ||
XM_015447285.1 | XP_015302771.1 | 5 - 6 | 1012 | 76 | ||
XM_015447286.1 | XP_015302772.1 | 6 - 7 | 713 | 76 | ||
XM_015447287.1 | XP_015302773.1 | 5 - 6 | 650 | 76 | ||
XM_015447288.1 | XP_015302774.1 | 5 - 6 | 648 | 76 | ||
XM_015447289.1 | XP_015302775.1 | 5 - 6 | 649 | 76 | ||
XM_015447290.1 | XP_015302776.1 | 5 - 6 | 711 | 76 | ||
XM_015447292.1 | XP_015302778.1 | 5 - 6 | 649 | 76 | ||
GenBank mRNA | AB168194.1 | - | 6 - 7 | 705 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |